Sodium hexafluorosilicate

Danger

250171-500G 250171-2KG
220048-166EA 81.54 USD
220048-166 220048-165
Sodium hexafluorosilicate
Sodium hexafluorosilicate
Formula: F₆Na₂Si
MW: 188.06 g/mol
Density: 2.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003491
CAS Number: 16893-85-9
EINECS: 240-934-8
UN: 2674
ADR: 6.1,III
Merck Index: 12,08769

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR