(S)-(+)-1-Bromo-2-methylbutane ≥99%

Danger

Synonyms: (S)-(+)-2-Methylbutyl bromide

250023-5G 250023-1G
220048-152EA 196 USD
220048-152 220048-151
(S)-(+)-1-Bromo-2-methylbutane ≥99%
(S)-(+)-1-Bromo-2-methylbutane
Formula: C₅H₁₁Br
MW: 151.05 g/mol
Boiling Pt: 122…123 °C (1013 hPa)
Density: 1.221 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078662
CAS Number: 534-00-9
EINECS: 208-583-5
UN: 1993
ADR: 3,II
Merck Index: 12,00642

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR