Tin(IV) chloride 1 M in dichloromethane

Synonyms: Stannic chloride, Tin tetrachloride

249955-800ML 249955-100ML
220048-147EA 141.23 USD
220048-147 220048-146
Tin(IV) chloride 1 M in dichloromethane
Tin(IV) chloride
Formula: Cl₄Sn
MW: 260.52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011242
CAS Number: 7646-78-8
EINECS: 231-588-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR