1-Bromo-3-methyl-2-butene ≥90%, Technical Grade

Synonyms: 3,3-Dimethylallyl bromide, 4-Bromo-2-methyl-2-butene

249904-25G 249904-5G
220048-142EA 107.37 USD
220048-142 220048-143
1-Bromo-3-methyl-2-butene ≥90%, Technical Grade
1-Bromo-3-methyl-2-butene
Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 129…133 °C (1013 hPa)
Density: 1.293 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000242
CAS Number: 870-63-3
EINECS: 212-799-5
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR