Methyl trifluoroacetate ≥99%

Synonyms: Trifluoroacetic methyl ester

249831-100G 249831-25G
220048-132EA 171.23 USD
220048-132 220048-133
Methyl trifluoroacetate ≥99%
Methyl trifluoroacetate
Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
Boiling Pt: 43 °C (1013 hPa)
Melting Pt: –78 °C
Density: 1.2639 g/cm³ (20 °C)
MDL Number: MFCD00000417
CAS Number: 431-47-0
EINECS: 207-074-5
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR