2-Naphthalenesulfonic acid, Technical Grade

Synonyms: Naphthalene-2-sulphonic acid, β-Naphthalenesulphonic acid

249548-5G 249548-100G
220048-084EA 26.92 USD
220048-084 220048-083
2-Naphthalenesulfonic acid, Technical Grade
2-Naphthalenesulphonic acid
Formula: C₁₀H₈O₃S
MW: 208.24 g/mol
Boiling Pt: ~ 350 °C (1013 hPa)
Melting Pt: 124 °C
Density: 1.441 g/cm³ (25 °C)
MDL Number: MFCD00149271
CAS Number: 120-18-3
EINECS: 204-375-3
Merck Index: 12,06464

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR