2,6-Difluorobenzoyl chloride ≥99%

249408-100G 249408-25G
220048-075EA 256.31 USD
220048-075 220048-076
2,6-Difluorobenzoyl chloride ≥99%
2,6-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Boiling Pt: 72 °C (1013 hPa)
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000659
CAS Number: 18063-02-0
EINECS: 241-971-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR