4-Amino-3-nitrophenol ≥98%

Synonyms: 4-Hydroxy-2-nitroaniline

249319-100G 249319-25G
220048-069EA 84.54 USD
220048-069 220048-070
4-Amino-3-nitrophenol ≥98%
4-Amino-3-nitrophenol
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 149 - 152 °C
MDL Number: MFCD00066310
CAS Number: 610-81-1
EINECS: 210-236-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR