1-Chloro-2-iodobenzene ≥99.0%

Warning

Synonyms: 2-Chloro-1-iodobenzene

24910-U 24910-5ML-F
220048-059EA 101.08 USD
220048-059 220048-058
1-Chloro-2-iodobenzene ≥99.0%
1-Chloro-2-iodobenzene
Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 1 °C
Density: 1.954 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001033
CAS Number: 615-41-8
EINECS: 210-423-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR