Triisopropylsilyl trifluoromethanesulfonate ≥97%

Synonyms: Trifluoromethanesulphonic acid triisopropylsilyl ester, Tri(propan-2-yl)silyl trifluoromethanesulfonate

248460-10G 248460-50G
220047-943EA 149.69 USD
220047-943 220047-944
Triisopropylsilyl trifluoromethanesulfonate ≥97%
Triisopropylsilyl trifluoromethanesulphonate
Formula: C₁₀H₂₁F₃O₃SSi
MW: 306.42 g/mol
Boiling Pt: 120 °C (1013 hPa)
Density: 1.136 g/cm³ (20 °C)
MDL Number: MFCD00009913
CAS Number: 80522-42-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR