3-Thenoic acid

Synonyms: 3-Thiophenecarboxylic acid, Thiophene-3-carboxylic acid

247766-1G 247766-5G
220047-897EA 37.69 USD
220047-897 220047-898
3-Thenoic acid
3-Thenoic acid
Formula: C₅H₄O₂S
MW: 128.15 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00005467
CAS Number: 88-13-1
EINECS: 201-802-5
Merck Index: 12,09415

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR