1,6-Diaminohexane dihydrochloride ≥99%

Synonyms: 1,6-Hexanediamine dihydrochloride, Hexane-1,6-diamine dihydrochloride, Hexamethylenediamine dihydrochloride

247731-100G 247731-25G
220047-895EA 182 USD
220047-895 220047-896
1,6-Diaminohexane dihydrochloride ≥99%
1,6-Diaminohexane dihydrochloride
Formula: C₆H₁₈Cl₂N₂
MW: 189.13 g/mol
Melting Pt: 258 °C
Density: 1.216 g/cm³ (20 °C)
MDL Number: MFCD00012528
CAS Number: 6055-52-3
EINECS: 227-977-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR