meso-Tetraphenylporphine

Synonyms: 5,10,15,20-Tetraphenyl-21H,23H-porphine, 5,10,15,20-Tetraphenylporphyrin, meso-Tetraphenylporphine, TPP

247367-250MG 247367-1G
220047-864EA 203 USD
220047-864 220047-863
meso-Tetraphenylporphine
meso-Tetraphenylporphine
Formula: C₄₄H₃₀N₄
MW: 614.75 g/mol
Density: 1.276 g/cm³ (20 °C)
MDL Number: MFCD00011680
CAS Number: 917-23-7
EINECS: 213-025-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9db3ad3f15ef176ad5c2987e5e320fa439882d5&vid=ec7727426cb2cadc4ec56eb17800889046284e1b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR