2-Acetyl-1H-pyrrole

Synonyms: Methyl-2-pyrrolylketon, Methyl 2-pyrrolyl ketone, 1-(1H-Pyrrol-2-yl)ethanone, 1-(1H-Pyrrol-2-yl)ethan-1-one, 2-Acetylpyrrole

247359-5G 247359-25G
220047-862EA 43.08 USD
220047-862 220047-861
2-Acetyl-1H-pyrrole
2-Acetyl-1H-pyrrole
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 63…65 °C
MDL Number: MFCD00005220
CAS Number: 1072-83-9
EINECS: 214-016-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR