6-Bromo-1-hexene ≥95%

Danger

247219-1G 247219-5G
220047-855EA 25.85 USD
220047-855 220047-856
6-Bromo-1-hexene ≥95%
6-Bromo-1-hexene
Formula: C₆H₁₁Br
MW: 163.06 g/mol
Boiling Pt: 34…36 °C (0.05 mmHg)
Density: 1.2169 g/cm³ (20 °C)
MDL Number: MFCD00000269
CAS Number: 2695-47-8
EINECS: 220-267-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR