5'-Fluoro-2'-hydroxyacetophenone ≥98%

Synonyms: 1-(5-Fluoro-2-hydroxyphenyl)ethanone

247170-5G 247170-1G
220047-851EA 32.42 USD
220047-851 220047-850
5'-Fluoro-2'-hydroxyacetophenone ≥98%
5'-Fluoro-2'-hydroxyacetophenone
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: 56 °C
MDL Number: MFCD00011668
CAS Number: 394-32-1
EINECS: 206-893-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR