3,4-Dibromothiophene

Warning

247154-25G 247154-5G
220047-847EA 75.17 USD
220047-847 220047-848
3,4-Dibromothiophene
3,4-Dibromothiophene
Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: 4.5 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005465
CAS Number: 3141-26-2
EINECS: 221-546-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR