2-Chlorothiophenol ≥99%

Synonyms: o-Chlorothiophenol, 2-chlorothiophenol, 1-Chloro-2-mercaptobenzene, 2-Chlorobenzenethiol, 2-Chlorobenzene-1-thiol

247146-25G 247146-5G
220047-845EA 81.2 USD
220047-845 220047-846
2-Chlorothiophenol ≥99%
2-Chlorothiophenol
Formula: C₆H₅ClS
MW: 144.62 g/mol
MDL Number: MFCD00004830
CAS Number: 6320-03-2
EINECS: 228-667-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR