2-Bromo-5-nitrotoluene ≥98%

Synonyms: 1-Bromo-2-methyl-4-nitrobenzene

247103-25G 247103-5G
220047-841EA 74.63 USD
220047-841 220047-842
2-Bromo-5-nitrotoluene ≥98%
2-Bromo-5-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 140 - 143 °C (17 -rr)
Melting Pt: 78 °C
MDL Number: MFCD00007281
CAS Number: 7149-70-4
EINECS: 230-481-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR