2-Acetyl-5-chlorothiophene

Synonyms: 1-(5-Chloro-2-thienyl)-1-ethanone, Rivaroxaban Impurity 20

247073-25G 247073-5G
220047-838EA 123.46 USD
220047-838 220047-839
2-Acetyl-5-chlorothiophene
2-Acetyl-5-chlorothiophene
Formula: C₆H₅ClOS
MW: 160.62 g/mol
Boiling Pt: 117 °C (17 -rr)
Melting Pt: 52 °C
MDL Number: MFCD00005444
CAS Number: 6310-09-4
EINECS: 228-630-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR