Tributylphosphine

Synonyms: Tri-n-butylphosphine, TBP

247049-100G 247049-500G
220047-786EA 74.77 USD
220047-786 220047-787
Tributylphosphine
Tributylphosphine
Formula: C₁₂H₂₇P
MW: 202.32 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.8165 g/cm³ (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00009462
CAS Number: 998-40-3
EINECS: 213-651-2
UN: 3254
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR