5-Sulfosalicylic acid dihydrate ≥99% ACS

Warning

Synonyms: 2-Hydroxy-5-sulphobenzoic acid dihydrate

247006-100G 247006-500G
220047-779EA 72.31 USD
220047-779 220047-780
5-Sulfosalicylic acid dihydrate ≥99% ACS
5-Sulphosalicylic acid dihydrate
Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
MW: 254.22 g/mol
Melting Pt: 224 °C
Density: 1.695 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00149540
CAS Number: 5965-83-3
EINECS: 202-555-6
UN: 2585
ADR: 8,III
Merck Index: 13,09054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69e00688850e885438e2fdd9a62f6d564ebf6ae8&vid=6a3840cf38254959a0062041e9a84fa6740b1288&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR