Ytterbium(III) oxide ≥99.9%

Synonyms: Diytterbium trioxide, Ytterbium oxide

246999-10G 246999-50G
220047-775EA 135.23 USD
220047-775 220047-776
Ytterbium(III) oxide ≥99.9%
Ytterbium(III) oxide
Formula: O₃Yb₂
MW: 394.08 g/mol
Melting Pt: 2346 °C
Density: 9.2 g/cm³ (20 °C)
MDL Number: MFCD00011290
CAS Number: 1314-37-0
EINECS: 215-234-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR