4-Phenoxybenzoic acid ≥97%

Warning

246182-5G 246182-25G
220047-689EA 428.08 USD
220047-689 220047-688
4-Phenoxybenzoic acid ≥97%
4-Phenoxybenzoic acid
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
MDL Number: MFCD00002539
CAS Number: 2215-77-2
EINECS: 218-682-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR