Ethyl isobutyrate ≥99%

Synonyms: 2-Methylpropionic acid ethyl ester

246085-500ML 246085-100ML
220046-990EA 66.88 USD
220046-990 220046-989
Ethyl isobutyrate ≥99%
Ethyl isobutyrate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 112 °C (1013 hPa)
Melting Pt: –88 °C
Density: 0.87 g/cm³ (20 °C)
MDL Number: MFCD00009165
CAS Number: 97-62-1
EINECS: 202-595-4
UN: 2385
ADR: 3,II
Merck Index: 13,03847

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR