Valeric anhydride ≥97%

Danger

Synonyms: Pentanoic anhydride

245933-100ML 245933-500ML
220047-675EA 136.77 USD
220047-675 220047-676
Valeric anhydride ≥97%
Valeric anhydride
Formula: C₁₀H₁₈O₃
MW: 186.25 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: -56 °C
Density: 0.94 g/cm³ (20 °C)
MDL Number: MFCD00009477
CAS Number: 2082-59-9
EINECS: 218-212-9
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR