2-Bromopyrimidine

Warning

245844-1G 245844-5G
220047-039EA 28.65 USD
220047-039 220047-040
2-Bromopyrimidine
2-Bromopyrimidine
Formula: C₄H₃BrN₂
Boiling Pt: 62 - 64 °C (1.5 -rr)
Melting Pt: 56 - 57.5 °C
Density: Min. 1 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00014601
CAS Number: 4595-60-2
EINECS: 224-993-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR