Diethyl adipate ≥99%, ReagentPlus®

Synonyms: NSC 19160, Adipic acid diethyl ester, 1,6-Hexanedioic acid diethyl ester, NSC 3363

245720-500G 245720-100G
220047-023EA 125.54 USD
220047-023 220047-022
Diethyl adipate ≥99%, ReagentPlus®
Diethyl adipate
Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 251 °C (lit.)
Melting Pt: −20…−19 °C (lit.)
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009215
CAS Number: 141-28-6
EINECS: 205-477-0
Merck Index: 13,00163

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR