3-Chlorophenyl isocyanate ≥99%

Synonyms: Isocyanic acid 3-Chlorophenyl ester, 3-Chlorophenylisocyanate

245682-100G 245682-5G
220047-546EA 199.23 USD
220047-546 220047-547
3-Chlorophenyl isocyanate ≥99%
3-Chlorophenyl isocyanate
Formula: C₇H₄ClNO
MW: 153.57 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: -4.4 °C
Density: 1.26 g/cm³ (20 °C)
MDL Number: MFCD00002016
CAS Number: 2909-38-8
EINECS: 220-822-5
UN: 3382
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR