Benzyl bromoacetate ≥96%

Synonyms: Benzyl 2-bromoacetate, Bromoacetic benzyl ester

245631-50G 245631-250G
220047-543EA 42.22 USD
220047-543 220047-542
Benzyl bromoacetate ≥96%
Benzyl bromoacetate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 130 - 135 °C (11 hPa)
Density: 1.446 g/cm³ (20 °C)
MDL Number: MFCD00000190
CAS Number: 5437-45-6
EINECS: 226-611-4

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR