ω-Bromo-4-nitroacetophenone ≥95%

Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone

245615-10G 245615-50G
220047-004EA 84.75 USD
220047-004 220047-005
ω-Bromo-4-nitroacetophenone ≥95%
ω-Bromo-4-nitroacetophenone
Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007356
CAS Number: 99-81-0
EINECS: 202-789-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR