4,4-Dimethyl-2-cyclohexen-1-one ≥97%

Synonyms: 4,4-Dimethylcyclohex-2-enone

245542-25G 245542-5G
220047-532EA 176.92 USD
220047-532 220047-533
4,4-Dimethyl-2-cyclohexen-1-one ≥97%
4,4-Dimethyl-2-cyclohexen-1-one
Formula: C₈H₁₂O
MW: 124.18 g/mol
Boiling Pt: 27.5 °C (0.2 -rr)
Density: 0.93 to 0.935 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009695
CAS Number: 1073-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR