Tetrakis(triphenylphosphine)platinum(0)

Synonyms: Tetrakis(triphenylphosphine)platinum

244961-1G 244961-5G
220047-475EA 157.54 USD
220047-475 220047-476
Tetrakis(triphenylphosphine)platinum(0)
Tetrakis(triphenylphosphine)platinum(0)
Formula: C₇₂H₆₀P₄Pt
MW: 1244.24 g/mol
MDL Number: MFCD00010014
CAS Number: 14221-02-4
EINECS: 238-087-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR