Methyldiphenylphosphine

244902-1G 244902-50G 244902-10G
220047-467EA 35.45 USD
220047-467 220047-468 220047-466
Methyldiphenylphosphine
Methyldiphenylphosphine
Formula: C₁₃H₁₃P
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.0784 g/cm³ (15 °C)
MDL Number: MFCD00008508
CAS Number: 1486-28-8
EINECS: 216-065-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR