Fentin hydride

Danger

Synonyms: Hydridotriphenylstannane, Triphenyltin hydride

244775-25G 244775-5G
220045-625EA 227.08 USD
220045-625 220045-626
Fentin hydride
Fentin hydride
Formula: C₁₈H₁₆Sn
MW: 351.04 g/mol
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: 27…28 °C
Density: 1.37 g/cm³ (20 °C)
MDL Number: MFCD00003004
CAS Number: 892-20-6
EINECS: 212-967-8
UN: 3146
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR