Lithium tetrafluoroborate ≥98%

244767-10G 244767-50G
220045-623EA 83.08 USD
220045-623 220045-624
Lithium tetrafluoroborate ≥98%
Lithium tetrafluoroborate
Formula: BF₄Li
MW: 93.74 g/mol
Density: 0.852 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011087
CAS Number: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR