1-(Trimethylsilyl)-1-propyne

Synonyms: Trimethyl-1-propynylsilane

244481-25G 244481-5G
220045-642EA 467.38 USD
220045-642 220045-643
1-(Trimethylsilyl)-1-propyne
1-(Trimethylsilyl)-1-propyne
Formula: C₆H₁₂Si
MW: 112.25 g/mol
Boiling Pt: 99 °C (1013 hPa)
Density: 0.759 g/cm³ (20 °C)
MDL Number: MFCD00009271
CAS Number: 6224-91-5
EINECS: 228-314-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR