3,3-Dimethyl-1-butyne ≥98%

Synonyms: 3,3-Dimethylbutyne

244392-25G 244392-5G
220047-417EA 407.08 USD
220047-417 220047-418
3,3-Dimethyl-1-butyne ≥98%
3,3-Dimethyl-1-butyne
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 38 °C (1013 hPa)
Melting Pt: –78.2 °C
Density: 1.0003 g/cm³ (0 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008852
CAS Number: 917-92-0
EINECS: 213-035-3
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR