2-Butyne-1-ol ≥98%

Danger

Synonyms: 2-Butyn-1-ol, But-2-yn-1-ol, 1-Hydroxy-2-butyne

244341-25G 244341-5G
220047-411EA 159.38 USD
220047-411 220047-412
2-Butyne-1-ol ≥98%
2-Butyne-1-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 142 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.9483 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002914
CAS Number: 764-01-2
EINECS: 212-113-4
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR