2-Iodobutane ≥99%

Synonyms: 2-Butyl iodide, sec-Butyl iodide

244325-100G 244325-5G
220047-409EA 107.26 USD
220047-409 220047-410
2-Iodobutane ≥99%
2-Iodobutane
Formula: C₄H₉I
MW: 184.02 g/mol
Boiling Pt: 119 °C (1013 hPa)
Melting Pt: -104 °C
Density: 1.6263 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001072
CAS Number: 513-48-4
EINECS: 208-163-1
UN: 2390
ADR: 3,II
Merck Index: 13,01573

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR