Nickel acetate tetrahydrate ≥98%

Danger

Synonyms: Nickel diacetate tetrahydrate, Nickel (II) acetate tetrahydrate, Nickel di(acetate) tetrahydrate, Acetic acid nickel (II) salt tetrahydrate

244066-100G 244066-500G
220046-972EA 52.31 USD
220046-972 220046-973
Nickel acetate tetrahydrate ≥98%
Nickel acetate tetrahydrate
Formula: C₄H₆NiO₄·4H₂O
MW: 248.84 g/mol
Boiling Pt: 16 °C (1013 hPa)
Density: 1.798 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066973
CAS Number: 6018-89-9
EINECS: 206-761-7
Merck Index: 12,06583

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR