Sodium molybdate(VI) ≥98%

Warning

Synonyms: Sodium molybdenum oxide, Sodium molybdate, Molybdic acid sodium salt, Molybdic acid, sodium salt, di-Sodium molybdate

243655-100G 243655-500G 243655-5G
220046-182EA 58.95 USD
220046-182 220046-233 220046-234
Sodium molybdate(VI) ≥98%
Sodium molybdate(VI)
Formula: MoNa₂O₄
MW: 205.92 g/mol
Melting Pt: 687 °C
Density: 3.28 g/cm³ (18 °C)
MDL Number: MFCD00003486
CAS Number: 7631-95-0
EINECS: 231-551-7
Merck Index: 13,08718

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR