Talc

Warning

243604-2.5KG 243604-500G
220047-359EA 609.23 USD
220047-359 220047-360
Talc
Talc
Formula: Mg₃O₁₁Si₄·1H₂O
Melting Pt: ? 1200 °C
Density: 2.58 to 2.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00792903
CAS Number: 14807-96-6
EINECS: 238-877-9
Merck Index: 13,09127

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR