Potassium perrhenate ≥99%

Danger

Synonyms: Potassium perrhenate (VII)

243590-25G 243590-5G
220047-356EA 1129.23 USD
220047-356 220047-357
Potassium perrhenate ≥99%
Potassium perrhenate
Formula: KO₄Re
MW: 289.3 g/mol
Boiling Pt: 1370 °C (1013 hPa)
Melting Pt: 550 °C
Density: 4.88 g/cm³ (20 °C)
MDL Number: MFCD00011365
CAS Number: 10466-65-6
EINECS: 233-953-8
UN: 1479
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR