Lead(II) carbonate basic

Danger

Synonyms: Lead carbonat basic, White lead, Lead (II) carbonate hydroxide, Lead (II) hydroxide carbonate

243582-2.5KG 243582-100G
220047-355EA 466.15 USD
220047-355 220047-354
Lead(II) carbonate basic
Lead(II) carbonate basic
Formula: (PbCO₃)₂·Pb(OH)₂
MW: 775.63 g/mol
Melting Pt: 400 °C
Density: 6.06 g/cm³ (20 °C)
MDL Number: MFCD00078155
CAS Number: 1319-46-6
EINECS: 215-290-6
UN: 2291
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR