Tellurium dioxide ≥99%

243450-50G 243450-250G
220047-336EA 264.62 USD
220047-336 220047-335
Tellurium dioxide ≥99%
Tellurium dioxide
Formula: O₂Te
MW: 159.6 g/mol
Boiling Pt: 1245 °C (1013 hPa)
Melting Pt: 773 °C
Density: 5.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,09123

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR