Strontium nitrate ≥99.0% ACS

Warning

243426-500G 243426-100G
220047-329EA 99.69 USD
220047-329 220047-328
Strontium nitrate ≥99.0% ACS
Strontium nitrate
Formula: Sr(NO₃)₂
MW: 211.63 g/mol
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 570 °C
Density: 2.113 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1,III
Merck Index: 13,08926

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR