Potassium bromide ≥99.0% ACS

Warning

243418-500G 243418-2.5KG 243418-100G
220047-327EA 67.69 USD
220047-327 220047-326 220047-325
Potassium bromide ≥99.0% ACS
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR