Chromium(III) potassium sulfate dodecahydrate ACS

Warning

Synonyms: Chromium potassium sulphate dodecahydrate, Chromium (III) potassium bis(sulphate) dodecahydrate

243361-100G 243361-500G 243361-5G
220047-315EA 51.02 USD
220047-315 220047-316 220047-317
Chromium(III) potassium sulfate dodecahydrate ACS
Chromium(III) potassium sulphate dodecahydrate
Formula: CrK(SO₄)₂·12H₂O
MW: 499.4 g/mol
Melting Pt: 89 °C
Density: 1.83 g/cm³ (20 °C)
MDL Number: MFCD00149917
CAS Number: 7788-99-0
EINECS: 233-401-6
Merck Index: 13,02250

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR