Allyl isocyanate ≥98%

Synonyms: 3-Isocyanatoprop-1-ene

243272-5G 243272-25G
220046-809EA 128.15 USD
220046-809 220046-808
Allyl isocyanate ≥98%
Allyl isocyanate
Formula: C₄H₅NO
MW: 83.09 g/mol
Boiling Pt: 87…89 °C (1013 hPa)
Density: 0.94 g/cm³ (20 °C)
MDL Number: MFCD00002044
CAS Number: 1476-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR